missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Palladium ICP Standard, 10,000 ppm Pd in 10% HCl, Ricca Chemical
Supplier: Ricca Chemical Company PPD10KH50
Specifications
Palladium ICP Standard | |
Natural Poly Bottle | |
67.0,23.73,0.67 | |
Brown | |
Pd | |
[Pd] | |
106.42 | |
CHEBI:33363 |
7440-05-3,7647-01-0,7732-18-5 | |
50 mL | |
70.0,24.7,0.7 | |
10,000ppm Pd | |
KDLHZDBZIXYQEI-UHFFFAOYSA-N | |
palladium | |
23938 | |
1 mL = 10 mg Pd |
Chemical Identifiers
7440-05-3 | |
106.42 | |
23938 | |
palladium |
Pd | |
KDLHZDBZIXYQEI-UHFFFAOYSA-N | |
CHEBI:33363 | |
[Pd] |