missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Palladium hydroxide, Pd 5% on carbon powder, Type A403002-5, nominally 50% water

Manufacturer:  Alfa Aesar™ 4557604

 View more versions of this product

Catalog No. AA4557604



Specifications

Specifications

Palladium hydroxide
12135-22-7
MFCD00064599
NXJCBFBQEVOTOW-UHFFFAOYSA-L
palladium(2+);dihydroxide
9942167
Odorless
2g
Pd 5% on carbon powder, Type A403002-5, Nominally 50% water
H2O2Pd
palladium hydroxide, palladium ii hydroxide, pearlman's catalyst, pearlmans catalyst, dihydroxypalladium, palladiumhydroxide, pearlman catalyst, peariman's catalyst, pearl man's catalyst, pd oh 2 on carbon
[OH-].[OH-].[Pd+2]
140.434
Powder
Black

Chemical Identifiers

12135-22-7
140.434
NXJCBFBQEVOTOW-UHFFFAOYSA-L
9942167
[OH-].[OH-].[Pd+2]
H2O2Pd
MFCD00064599
palladium hydroxide, palladium ii hydroxide, pearlman's catalyst, pearlmans catalyst, dihydroxypalladium, palladiumhydroxide, pearlman catalyst, peariman's catalyst, pearl man's catalyst, pd oh 2 on carbon
palladium(2+);dihydroxide
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS Documents