missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Palladium hydroxide, Pd 5% on carbon powder, Type A403002-5, nominally 50% water, Thermo Scientific Chemicals
$296.66 - $1269.81
Chemical Identifiers
CAS | 12135-22-7 |
---|---|
Molecular Formula | H2O2Pd |
Molecular Weight (g/mol) | 140.43 |
MDL Number | MFCD00064599 |
InChI Key | NXJCBFBQEVOTOW-UHFFFAOYSA-L |
Synonym | palladium hydroxide, palladium ii hydroxide, pearlman's catalyst, pearlmans catalyst, dihydroxypalladium, palladiumhydroxide, pearlman catalyst, peariman's catalyst, pearl man's catalyst, pd oh 2 on carbon |
PubChem CID | 9942167 |
IUPAC Name | palladium(2+);dihydroxide |
SMILES | [OH-].[OH-].[Pd++] |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
12135-22-7 | |
140.43 | |
NXJCBFBQEVOTOW-UHFFFAOYSA-L | |
9942167 | |
[OH-].[OH-].[Pd++] |
H2O2Pd | |
MFCD00064599 | |
palladium hydroxide, palladium ii hydroxide, pearlman's catalyst, pearlmans catalyst, dihydroxypalladium, palladiumhydroxide, pearlman catalyst, peariman's catalyst, pearl man's catalyst, pd oh 2 on carbon | |
palladium(2+);dihydroxide |
Specifications
Black | |
Powder | |
H2O2Pd | |
palladium hydroxide, palladium ii hydroxide, pearlman's catalyst, pearlmans catalyst, dihydroxypalladium, palladiumhydroxide, pearlman catalyst, peariman's catalyst, pearl man's catalyst, pd oh 2 on carbon | |
[OH-].[OH-].[Pd++] | |
140.43 | |
Odorless |
12135-22-7 | |
2 g | |
MFCD00064599 | |
NXJCBFBQEVOTOW-UHFFFAOYSA-L | |
palladium(2+);dihydroxide | |
9942167 | |
Palladium hydroxide, Pd 5% on carbon powder, Type A403002-5, Nominally 50% water |
Safety and Handling
P210-P240-P241-P280-P370+P378q
H228
EINECSNumber : 235-219-2
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only