Learn More
Palladium hydroxide, Pd 20% on carbon, nominally 50% water, Pearlman's Catalyst, Thermo Scientific Chemicals
$216.51 - $216.51
Chemical Identifiers
CAS | 12135-22-7 |
---|---|
Molecular Formula | H2O2Pd |
Molecular Weight (g/mol) | 140.43 |
MDL Number | MFCD00064599 |
InChI Key | NXJCBFBQEVOTOW-UHFFFAOYSA-L |
Synonym | palladium hydroxide, palladium ii hydroxide, pearlman's catalyst, pearlmans catalyst, dihydroxypalladium, palladiumhydroxide, pearlman catalyst, peariman's catalyst, pearl man's catalyst, pd oh 2 on carbon |
PubChem CID | 9942167 |
IUPAC Name | palladium(2+);dihydroxide |
SMILES | [OH-].[OH-].[Pd++] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA4257804
|
Thermo Scientific Chemicals
04257804 |
2 g |
Each for $216.51
|
|
|||||
Description
Hydrogenolysis of benzyl-nitrogen bondsPalladium hydroxide, Pd 20% on carbon, nominally 50% water is used as a catalyst for hydrogenolysis of benzyl groups and epoxides. It is used to reduce nitro compounds to amines. It acts as a dehydrogenation reagent as well as a useful reagent for debenzylation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Used as catalyst for hydrogenolysis of benzyl groups and epoxides.It is used to reduce nitro compounds to amines.
Acts as dehydrogenation reagent as well as useful reagent for debenzylation
Chemical Identifiers
12135-22-7 | |
140.43 | |
NXJCBFBQEVOTOW-UHFFFAOYSA-L | |
9942167 | |
[OH-].[OH-].[Pd++] |
H2O2Pd | |
MFCD00064599 | |
palladium hydroxide, palladium ii hydroxide, pearlman's catalyst, pearlmans catalyst, dihydroxypalladium, palladiumhydroxide, pearlman catalyst, peariman's catalyst, pearl man's catalyst, pd oh 2 on carbon | |
palladium(2+);dihydroxide |
Specifications
12135-22-7 | |
2 g | |
MFCD00064599 | |
Insoluble in water. | |
[OH-].[OH-].[Pd++] | |
140.43 | |
Palladium Hydroxide, 20% on Carbon, nominally 50% water |
Moist Powder | |
H2O2Pd | |
palladium hydroxide, palladium ii hydroxide, pearlman's catalyst, pearlmans catalyst, dihydroxypalladium, palladiumhydroxide, pearlman catalyst, peariman's catalyst, pearl man's catalyst, pd oh 2 on carbon | |
NXJCBFBQEVOTOW-UHFFFAOYSA-L | |
palladium(2+);dihydroxide | |
9942167 |
Safety and Handling
P210-P240-P241-P280-P370+P378q
H228
EINECSNumber : 235-219-2
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only