missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Palladium Hydroxide (contains Pd, PdO) on Carbon (wetted with ca. 50% Water), TCI America™

Manufacturer:  TCI America P152850G

 View more versions of this product

Catalog No. P152850G



Specifications

Specifications

Palladium Hydroxide (contains Pd, PdO) on Carbon (wetted with ca. 50% Water)
H2O2Pd
Pearlman′Cs Catalyst (contains Pd, PdO)
[OH-].[OH-].[Pd+2]
140.434
140.43
Black
12135-22-7
MFCD00064599
NXJCBFBQEVOTOW-UHFFFAOYSA-L
palladium(2+);dihydroxide
9942167
Crystal-Powder at 20°C
50g

Chemical Identifiers

12135-22-7
140.434
NXJCBFBQEVOTOW-UHFFFAOYSA-L
9942167
[OH-].[OH-].[Pd+2]
H2O2Pd
MFCD00064599
Pearlman′Cs Catalyst (contains Pd, PdO)
palladium(2+);dihydroxide
Safety and Handling

Safety and Handling

EINECSNumber : (1)-1141

TSCA : Yes

Documents