Please login to your online account to display your discounted pricing

Palladium Granules, 99.8% (Metals basis), Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Palladium
CAS 7440-05-3
Physical Form Granular
Assay Percent Range 99.8% (Metals basis)
MDL Number MFCD00011167
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1223703

  SDS
alfa aesar
1223703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1223706

  SDS
alfa aesar
1223706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1223714

  SDS
alfa aesar
1223714
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 1g
Color Silver-gray
Physical Form Granular
Chemical Name or Material Palladium
Assay Percent Range 99.8% (Metals basis)
MDL Number MFCD00011167
CAS 7440-05-3
Odor Odorless
Appearance Silver gray granules