missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Palladium Granules, 99.8% (Metals basis), Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific 1223714

 View more versions of this product

Catalog No. AA1223714



Specifications

Specifications

Palladium
7440-05-3
99.8% (Metals basis)
MFCD00011167
KDLHZDBZIXYQEI-UHFFFAOYSA-N
palladium
106.42
CHEBI:33363
Silver to Gray
25g
Silver gray granules
Pd
black
[Pd]
Odorless
23938
Granular

Chemical Identifiers

7440-05-3
106.42
KDLHZDBZIXYQEI-UHFFFAOYSA-N
23938
palladium
Pd
MFCD00011167
black
CHEBI:33363
[Pd]
Safety and Handling

Safety and Handling

EINECSNumber : 231-115-6

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents