Please login to your online account to display your discounted pricing

Alfa Aesar™ Palladium foil, 1.0mm (0.04 in.) thick, Premion™, 99.99% (metals basis)

Chemical Identifiers

Chemical Name or Material Palladium foil
CAS 7440-05-3
Name Note 1.0mm (0.04 in.) thick
Purity Grade Premion™
MDL Number MFCD00011167
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA12056FF

 
alfa aesar™
12056FF
25x25mm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA12056FI

 
alfa aesar™
12056FI
50x50mm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Palladium foil
TSCA TSCA
Name Note 1.0mm (0.04 in.) thick
Purity Grade Premion™
Physical Form Foil
Assay 99.99%
Assay Percent Notes (metals basis)
MDL Number MFCD00011167
Odor Odorless
Weight ≈7.5g/25x25mm
Quantity 25x25mm
CAS 7440-05-3