Please login to your online account to display your discounted pricing

Alfa Aesar™ Palladium foil, 1.0mm (0.04 in.) thick, 99.9% (metals basis)

Chemical Identifiers

Chemical Name or Material Palladium foil
CAS 7440-05-3
Name Note 1.0mm (0.04 in.) thick
MDL Number MFCD00011167
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA11513FF

  SDS
alfa aesar™
11513FF
25x25mm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA11513FI

  SDS
alfa aesar™
11513FI
50x50mm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Palladium foil
TSCA TSCA
Name Note 1.0mm (0.04 in.) thick
Physical Form Foil
Assay 99.9%
Assay Percent Notes (metals basis)
MDL Number MFCD00011167
CAS 7440-05-3
Odor Odorless
Weight ≈7.5g/25x25mm
Quantity 25x25mm