Please login to your online account to display your discounted pricing

Alfa Aesar™ Palladium foil, 0.5mm (0.02 in.) thick, 99.95+% (metals basis)

Chemical Identifiers

Chemical Name or Material Palladium foil
CAS 7440-05-3
Name Note 0.5mm (0.02 in.) thick
MDL Number MFCD00011167
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA11917FF

  SDS
alfa aesar™
11917FF
25x25mm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA11917FI

  SDS
alfa aesar™
11917FI
50x50mm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Name Note 0.5mm (0.02 in.) thick
Quantity 25x25mm
Chemical Name or Material Palladium foil
Weight ≈3.8g/25x25mm
MDL Number MFCD00011167
Physical Form Foil
CAS 7440-05-3
Odor Odorless
TSCA TSCA
Assay ≥99.95%
Assay Percent Notes (metals basis)