Please login to your online account to display your discounted pricing

Alfa Aesar™ Palladium foil, 0.5mm (0.02 in.) thick, 99.9% (metals basis)

Chemical Identifiers

Chemical Name or Material Palladium foil
CAS 7440-05-3
Name Note 0.5mm (0.02 in.) thick
MDL Number MFCD00011167
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA11514FF

 
alfa aesar™
11514FF
25x25mm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA11514FI

 
alfa aesar™
11514FI
50x50mm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Palladium foil
TSCA TSCA
Name Note 0.5mm (0.02 in.) thick
Physical Form Foil
Assay 99.9%
Assay Percent Notes (metals basis)
MDL Number MFCD00011167
CAS 7440-05-3
Odor Odorless
Weight ≈3.8g/25x25mm
Quantity 25x25mm