Please login to your online account to display your discounted pricing

Alfa Aesar™ Palladium foil, 0.25mm (0.01 in.) thick, 99.98+% (metals basis)

Chemical Identifiers

Chemical Name or Material Palladium foil
CAS 7440-05-3
Name Note 0.25mm (0.01 in.) thick
MDL Number MFCD00011167
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA11918FF

  SDS
alfa aesar™
11918FF
25x25mm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA11918FI

  SDS
alfa aesar™
11918FI
50x50mm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Odor Odorless
CAS 7440-05-3
Assay ≥99.98%
Assay Percent Notes (metals basis)
Weight ≈1.9g/25x25mm
TSCA TSCA
Chemical Name or Material Palladium foil
MDL Number MFCD00011167
Quantity 25x25mm
Physical Form Foil
Name Note 0.25mm (0.01 in.) thick