missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Palladium foil, 0.25mm (0.01 in.) thick, 99.98+% (metals basis)

Manufacturer:  Alfa Aesar™ 11918FI

 View more versions of this product

Catalog No. AA11918FI



Specifications

Specifications

Palladium foil
50 x 50mm
7440-05-3
Pd
black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon
[Pd]
Odorless
23938
Foil
≈1.9g/25x25mm
0.25mm (0.01 in.) thick
(metals basis)
MFCD00011167
KDLHZDBZIXYQEI-UHFFFAOYSA-N
palladium
106.42
CHEBI:33363
≥99.98%

Chemical Identifiers

7440-05-3
106.42
KDLHZDBZIXYQEI-UHFFFAOYSA-N
23938
palladium
Pd
MFCD00011167
black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon
CHEBI:33363
[Pd]
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents