Please login to your online account to display your discounted pricing

Alfa Aesar™ Palladium foil, 0.25mm (0.01 in.) thick, 99.9% (metals basis)

Chemical Identifiers

Chemical Name or Material Palladium foil
CAS 7440-05-3
Name Note 0.25mm (0.01 in.) thick
MDL Number MFCD00011167
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA11516FF

  SDS
alfa aesar™
11516FF
25x25mm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA11516FI

  SDS
alfa aesar™
11516FI
50x50mm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA11516GH

  SDS
alfa aesar™
11516GH
100x100mm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Odor Odorless
TSCA TSCA
Assay Percent Notes (metals basis)
Chemical Name or Material Palladium foil
MDL Number MFCD00011167
Physical Form Foil
CAS 7440-05-3
Assay 99.9%
Weight ≈1.9g/25x25mm
Name Note 0.25mm (0.01 in.) thick
Quantity 25x25mm