Please login to your online account to display your discounted pricing

Palladium on calcium carbonate, reduced 5% Pd, ACROS Organics™

$555.09 - $555.09

Chemical Identifiers

Chemical Name or Material Palladium on calcium carbonate
CAS 2023568
CAS 471-34-1
Molecular Formula Pd
Formula Weight 106.42
MDL Number MFCD00151707
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC388980500

  SDS
thermo fisher scientific
AC388980500
Glass bottle 50g Each for $555.09
Description & Specifications

Specifications

Formula Weight 106.42
Physical Form Powder
Quantity 50g
Color Gray
Water 5% Max.
Merck Index 15, 7090
CAS 2023568
CAS 471-34-1
Chemical Name or Material Palladium on calcium carbonate
MDL Number MFCD00151707
Packaging Glass bottle
Palladium (Pd) 5%, exact Pd-content on the certificate of analysis
Molecular Formula Pd
Health Hazard 3 GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.