Please login to your online account to display your discounted pricing

Palladium on barium sulfate, 5% Pd, unreduced, ACROS Organics™


Quantity 50g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  195060500

 View more versions of this product

Catalog No. AC195060500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material Palladium on barium sulfate
Quantity 50g
Name Note 5% Pd
CAS 2023568
RTECS Number RT3480500
EINECS Number 231-115-6
TSCA TSCA
Physical Form Powder
Assay 4.50 to 5.50% (Pd)
Molecular Formula Pd
Formula Weight 106.42
MDL Number MFCD03613605
Fieser 01,778; 04,365; 05,498; 06,443; 07,274; 15,13; 16,356
Merck Index 15, 7090
Packaging Glass bottle
Water 5% Max. (K.F.)
Color Brown

Specifications

Chemical Name or Material Palladium on barium sulfate
Name Note 5% Pd
CAS 2023568
Assay 4.50 to 5.50% (Pd)
Molecular Formula Pd
Formula Weight 106.42
MDL Number MFCD03613605