Please login to your online account to display your discounted pricing

Palladium, Alfa Aesar™

Form: 900-1100m2;/g; Sold on a dry weight basis. Unit weight excludes water weight

Chemical Identifiers

Chemical Name or Material Palladium
Name Note 10% on activated carbon powder
Physical Form Powder
MDL Number MFCD03457879
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3830406

  SDS
alfa aesar
3830406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3830414

  SDS
alfa aesar
3830414
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 5g
Color Black
Physical Form Powder
Chemical Name or Material Palladium
Appearance Moist black powder
Name Note 10% on activated carbon powder
MDL Number MFCD03457879