Please login to your online account to display your discounted pricing

Palladium on activated carbon, unreduced, 5% Pd, ACROS Organics™


Quantity 500g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC195025000

 View more versions of this product

Catalog No. AC195025000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 106.42
Assay 4.8 to 5.2% (Pd, on dry basis)
Physical Form Crystals or Crystalline Powder and/or Chunks
Quantity 500g
Color White
Merck Index 15, 7090
Particle Size Typicaly 10% (<4 Micron) , 50% (<21 Micron), 90% (<90 Micron)
Fieser 01,778; 04,365; 05,498; 06,443; 07,274; 15,13; 16,356
Health Hazard 1 Warning
Name Note 5% Pd
MDL Number MFCD03457879
CAS 7440-05-3
Chemical Name or Material Palladium on activated carbon
Molecular Formula Pd
Loss on Drying 3 % max. (120°C)
Packaging Glass bottle
CAS 7440-44-0
Health Hazard 3 GHS P Statement:
Keep container tightly closed.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Flammable solid.

Specifications

Chemical Name or Material Palladium on activated carbon
Name Note 5% Pd
CAS 7440-05-3
CAS 7440-44-0
Assay 4.8 to 5.2% (Pd, on dry basis)
Molecular Formula Pd
Formula Weight 106.42
MDL Number MFCD03457879
Accessories

Warning

GHS P Statement:
Keep container tightly closed.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Flammable solid.

SDS & Safety Documents