Please login to your online account to display your discounted pricing

Palladium on activated carbon, 3% Pd, unreduced, ACROS Organics™


Quantity 10g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  217870100

 View more versions of this product

Catalog No. AC217870100

  • / Each
 

Description & Specifications

Chemical Identifiers

Chemical Name or Material Palladium on activated carbon
Quantity 10g
Name Note 3% Pd
CAS 7440-05-3
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement:
Flammable solid.
CAS 7440-44-0
Health Hazard 3 GHS P Statement:
Keep container tightly closed.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Physical Form Powder
Assay 3% (Pd)
Molecular Formula Pd
Formula Weight 106.42
MDL Number MFCD03457879
Fieser 01,778; 04,365; 05,498; 06,443; 07,274; 15,13; 16,356
Merck Index 15, 7090
Packaging Glass bottle
Particle Size Typical particle size distribution: D50: 20 Micron, < 8 Micron: 17%, <16 Micron: 39%, <32 Micron: 67%, <64Micron: 90%, <128 Micron: 99%
Pore Volume total pore volume (ml/g): 1.00
Specific Surface Area 1000m2/g
Color Black

Specifications

Chemical Name or Material Palladium on activated carbon
Name Note 3% Pd
CAS 7440-05-3
CAS 7440-44-0
Assay 3% (Pd)
Molecular Formula Pd
Formula Weight 106.42
MDL Number MFCD03457879
Accessories

Warning

GHS P Statement:
Keep container tightly closed.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Flammable solid.

SDS & Safety Documents