Please login to your online account to display your discounted pricing

Palladium, 99.99%, (trace metal basis), granules, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Palladium
CAS 2023568
Purity Grade For trace metal basis analysis
Assay Percent Range 100.00%
Molecular Formula Pd
Linear Formula Pd
Formula Weight 106.4g/mol
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC437020010

  SDS
thermo fisher scientific
437020010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Packaging Glass bottle
Total Trace Metal Impurities 50ppm max.
Color Gray
Quantity 1g
Chemical Name or Material Palladium
CAS 2023568
Purity Grade For trace metal basis analysis
Physical Form Gray crystalline granules
Assay Percent Range 100.00%
Molecular Formula Pd
Linear Formula Pd
Formula Weight 106.4g/mol
Health Hazard 1

for ultra trace analysis

Safety and Handling