Please login to your online account to display your discounted pricing

Palladium, 99.99%, (trace metal basis), granules, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC437020010

 View more versions of this product

Catalog No. AC437020010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystalline Granules
Formula Weight 106.4
Quantity 1g
Color Gray
Purity Grade For trace metal basis analysis
CAS 2023568
Chemical Name or Material Palladium
Assay Percent Range 100.00%
Molecular Formula Pd
Linear Formula Pd
Packaging Glass bottle
Health Hazard 1
Total Trace Metal Impurities 50ppm max.

Specifications

Chemical Name or Material Palladium
CAS 2023568
Purity Grade For trace metal basis analysis
Assay Percent Range 100.00%
Molecular Formula Pd
Linear Formula Pd
Formula Weight 106.4
Accessories

SDS & Safety Documents