Please login to your online account to display your discounted pricing

Alfa Aesar™ Palladium, 5% on strontium carbonate powder, reduced

Chemical Identifiers

Chemical Name or Material Palladium
Name Note 5% on strontium carbonate powder, Reduced
MDL Number MFCD00192595
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3981904

  SDS
alfa aesar™
3981904
2g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3981909

  SDS
alfa aesar™
3981909
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3981918

  SDS
alfa aesar™
3981918
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Palladium
TSCA TSCA
Name Note 5% on strontium carbonate powder, Reduced
MDL Number MFCD00192595
Quantity 2g