missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Palladium, 5% on gamma alumina powder, reduced, Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific 1171322

 View more versions of this product

Catalog No. AA1171322



Specifications

Specifications

Palladium
5% on gamma alumina
Dark gray powder
MFCD03613602
Insoluble in water.
[Pd]
Odorless
23938
Dark grey powder
100g
7440-05-3
Pd
black, charcoal, element, on carbon, on charcoal, paladio, palladex 600, palladium, element, palladium-activated carbon, palladium/carbon
KDLHZDBZIXYQEI-UHFFFAOYSA-N
palladium
106.42
CHEBI:33363
Gray

Chemical Identifiers

7440-05-3
106.42
KDLHZDBZIXYQEI-UHFFFAOYSA-N
23938
palladium
Pd
MFCD03613602
black, charcoal, element, on carbon, on charcoal, paladio, palladex 600, palladium, element, palladium-activated carbon, palladium/carbon
CHEBI:33363
[Pd]
Safety and Handling

Safety and Handling

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents