Please login to your online account to display your discounted pricing

Alfa Aesar™ Palladium, 5% on carbon paste, Type 761

Chemical Identifiers

Chemical Name or Material Palladium
CAS 7440-05-3
Name Note 5% on carbon paste, Type 761
MDL Number MFCD03457879
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3752606

 
alfa aesar™
H3752606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH3752614

 
alfa aesar™
H3752614
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH3752622

 
alfa aesar™
H3752622
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Palladium
TSCA TSCA
Odor Odorless
Name Note 5% on carbon paste, Type 761
Physical Form Solid
MDL Number MFCD03457879
CAS 7440-05-3
Color Dark gray
Quantity 5g