Please login to your online account to display your discounted pricing

Alfa Aesar™ Palladium, 5% on carbon paste, sulfided, Type A103023-5

Chemical Identifiers

Chemical Name or Material Palladium
CAS 7440-05-3
Name Note 5% on carbon paste, sulfided, Type A103023-5
MDL Number MFCD03457879
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3740006

  SDS
alfa aesar™
H3740006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH3740014

  SDS
alfa aesar™
H3740014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH3740022

  SDS
alfa aesar™
H3740022
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Palladium
Name Note 5% on carbon paste, sulfided, Type A103023-5
CAS 7440-05-3
TSCA TSCA
Physical Form Powder
MDL Number MFCD03457879
Odor Odorless
Color Black
Quantity 5g