missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Palladium, 5% on calcium carbonate powder, reduced

Manufacturer:  Alfa Aesar™ 1172306

 View more versions of this product

Catalog No. AA1172306

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Palladium
7440-05-3
MFCD03427452
KDLHZDBZIXYQEI-UHFFFAOYSA-N
palladium
23938
Odorless
5% on calcium carbonate powder, Reduced
Pd
black
[Pd]
106.42
CHEBI:33363
5g

Chemical Identifiers

7440-05-3
106.42
KDLHZDBZIXYQEI-UHFFFAOYSA-N
23938
palladium
Pd
MFCD03427452
black
CHEBI:33363
[Pd]
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents