missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Palladium, 5% on calcium carbonate powder, reduced

Chemical Identifiers

CAS 7440-05-3
Molecular Formula Pd
Molecular Weight (g/mol) 106.42
MDL Number MFCD03427452
InChI Key KDLHZDBZIXYQEI-UHFFFAOYSA-N
Synonym black, charcoal, element, on carbon, on charcoal, paladio, palladex 600, palladium, element, palladium-activated carbon, palladium/carbon
PubChem CID 23938
ChEBI CHEBI:33363
IUPAC Name palladium
SMILES [Pd]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1172306
View Documents
Alfa Aesar™
1172306
5g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AA1172314
View Documents
Alfa Aesar™
1172314
25g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AA1172322
View Documents
Alfa Aesar™
1172322
100g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

7440-05-3
106.42
KDLHZDBZIXYQEI-UHFFFAOYSA-N
23938
palladium
Pd
MFCD03427452
black, charcoal, element, on carbon, on charcoal, paladio, palladex 600, palladium, element, palladium-activated carbon, palladium/carbon
CHEBI:33363
[Pd]

Specifications

7440-05-3
MFCD03427452
Insoluble in most organic solvents.
[Pd]
106.42
CHEBI:33363
5g
Pd
black, charcoal, element, on carbon, on charcoal, paladio, palladex 600, palladium, element, palladium-activated carbon, palladium/carbon
KDLHZDBZIXYQEI-UHFFFAOYSA-N
palladium
23938
Odorless
Palladium, 5% on calcium carbonate powder, Reduced
Safety and Handling

Safety and Handling

TSCA : Yes

Recommended Storage : Ambient temperatures