missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Palladium, 5% on calcium carbonate powder, reduced

Manufacturer:  Alfa Aesar™ 1172322

 View more versions of this product

Catalog No. AA1172322

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

Palladium
7440-05-3
MFCD03427452
Insoluble in most organic solvents.
[Pd]
106.42
CHEBI:33363
100g
5% on calcium carbonate powder, Reduced
Pd
black, charcoal, element, on carbon, on charcoal, paladio, palladex 600, palladium, element, palladium-activated carbon, palladium/carbon
KDLHZDBZIXYQEI-UHFFFAOYSA-N
palladium
23938
Odorless

Chemical Identifiers

7440-05-3
106.42
KDLHZDBZIXYQEI-UHFFFAOYSA-N
23938
palladium
Pd
MFCD03427452
black, charcoal, element, on carbon, on charcoal, paladio, palladex 600, palladium, element, palladium-activated carbon, palladium/carbon
CHEBI:33363
[Pd]
Safety and Handling

Safety and Handling

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents