Please login to your online account to display your discounted pricing

Alfa Aesar™ Palladium, 5% on calcium carbonate, A303060-5

Chemical Identifiers

Chemical Name or Material Palladium
CAS 7440-05-3
Name Note 5% on calcium carbonate, A303060-5
MDL Number MFCD03427452
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3662214

  SDS
alfa aesar™
H3662214
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH3662206

  SDS
alfa aesar™
H3662206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH3662222

  SDS
alfa aesar™
H3662222
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Palladium
Name Note 5% on calcium carbonate, A303060-5
CAS 7440-05-3
TSCA TSCA
Physical Form Powder
MDL Number MFCD03427452
Odor Odorless
Color Dark brown or gray to black
Quantity 25g