missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Palladium, 5% on barium sulfate powder, unreduced

Hydrodehalogenation, Rosenmund reduction, asymetric hydrogenation

Manufacturer:  Alfa Aesar™ 2116209

 View more versions of this product

Catalog No. AA2116209



Specifications

Specifications

Palladium
7440-05-3
MFCD03613605
KDLHZDBZIXYQEI-UHFFFAOYSA-N
palladium
23938
Brown
5% on barium sulfate powder, Unreduced
Pd
black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon
[Pd]
106.42
CHEBI:33363
10g

Chemical Identifiers

7440-05-3
106.42
KDLHZDBZIXYQEI-UHFFFAOYSA-N
23938
palladium
Pd
MFCD03613605
black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon
CHEBI:33363
[Pd]
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS Documents