Please login to your online account to display your discounted pricing

Alfa Aesar™ Palladium, 5% on barium sulfate powder, unreduced

Hydrodehalogenation, Rosenmund reduction, asymetric hydrogenation

$108.97 - $108.97

Chemical Identifiers

Chemical Name or Material Palladium
Name Note 5% on barium sulfate powder, Unreduced
MDL Number MFCD03613605
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA2116209

 
alfa aesar™
2116209
10g Each for $108.97

AA2116218

 
alfa aesar™
2116218
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Palladium
TSCA TSCA
Name Note 5% on barium sulfate powder, Unreduced
MDL Number MFCD03613605
Color Pale brown
Quantity 10g