Please login to your online account to display your discounted pricing

Alfa Aesar™ Palladium, 5% on alumina powder, A302099-5

Chemical Identifiers

Chemical Name or Material Palladium
CAS 7440-05-3
Name Note 5% on alumina powder, A302099-5
MDL Number MFCD03613602
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3736806

  SDS
alfa aesar™
H3736806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH3736814

  SDS
alfa aesar™
H3736814
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH3736822

  SDS
alfa aesar™
H3736822
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Palladium
Name Note 5% on alumina powder, A302099-5
CAS 7440-05-3
TSCA TSCA
Physical Form Solid
MDL Number MFCD03613602
Odor Odorless
Color Dark gray
Quantity 5g