Please login to your online account to display your discounted pricing

Alfa Aesar™ Palladium, 5% on activated carbon powder, Type A401102-5, eggshell unreduced, nominally 50% water wet

Aromatic nitro group hydrogenation

Chemical Identifiers

Chemical Name or Material Palladium
Name Note 5% on activated carbon powder, Type A401102-5, eggshell unreduced, nominally 50% water wet
MDL Number MFCD03457879
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3829906

  SDS
alfa aesar™
3829906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3829914

  SDS
alfa aesar™
3829914
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3829922

  SDS
alfa aesar™
3829922
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Palladium
TSCA TSCA
Name Note 5% on activated carbon powder, Type A401102-5, eggshell unreduced, nominally 50% water wet
MDL Number MFCD03457879
Color Black
Quantity 5g
Odor Odorless
Weight 900 to 1100m2/g