Please login to your online account to display your discounted pricing

Alfa Aesar™ Palladium, 5% on activated carbon powder, Type A109047-5, standard, reduced, nominally 50% water wet

Chemical Identifiers

Chemical Name or Material Palladium
Name Note 5% on activated carbon powder, Type A109047-5, standard, reduced, nominally 50% water wet
MDL Number MFCD03457879
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4701914

 
alfa aesar™
4701914
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Palladium
TSCA TSCA
Odor Odorless
Name Note 5% on activated carbon powder, Type A109047-5, standard, reduced, nominally 50% water wet
Physical Form Powder
MDL Number MFCD03457879
Color Black
Quantity 25g