Please login to your online account to display your discounted pricing

Alfa Aesar™ Palladium, 5% on activated carbon powder, Type 487

Chemical Identifiers

Chemical Name or Material Palladium
CAS 7440-05-3
Name Note 5% on activated carbon powder, Type 487
MDL Number MFCD03457879
UN Number UN1362
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3758706

  SDS
alfa aesar™
H3758706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH3758714

  SDS
alfa aesar™
H3758714
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH3758722

  SDS
alfa aesar™
H3758722
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Palladium
Name Note 5% on activated carbon powder, Type 487
CAS 7440-05-3
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H228-H252
Flammable solid.
Self-heating in large quantities
may catch fire.
Health Hazard 3 GHS P Statement
P210-P280-P235+P410-P240-P370+P378a-P420
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Keep cool.
Protect from sunlight.
Ground/bond container and receiving equipment.
In case of fire: Use for extinction: CO2, powder or water spray.
Store away from other materials.
DOT Information Transport Hazard Class: 4.2; Packaging Group: III; Proper Shipping Name: CARBON, ACTIVATED
TSCA TSCA
Physical Form Powder
MDL Number MFCD03457879
UN Number UN1362
Odor Odorless
Color Dark gray
Quantity 5g
Safety and Handling

Danger

GHS P Statement
P210-P280-P235+P410-P240-P370+P378a-P420
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Keep cool.
Protect from sunlight.
Ground/bond container and receiving equipment.
In case of fire: Use for extinction: CO2, powder or water spray.
Store away from other materials.

GHS H Statement
H228-H252
Flammable solid.
Self-heating in large quantities
may catch fire.