Please login to your online account to display your discounted pricing

Alfa Aesar™ Palladium, 5% on activated carbon paste, Type 424

Chemical Identifiers

Chemical Name or Material Palladium
CAS 7440-05-3
Name Note 5% on activated carbon paste, Type 424
MDL Number MFCD03457879
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3742906

  SDS
alfa aesar™
H3742906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH3742914

  SDS
alfa aesar™
H3742914
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH3742922

  SDS
alfa aesar™
H3742922
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Name Note 5% on activated carbon paste, Type 424
Quantity 5g
Color Dark gray
Odor Odorless
CAS 7440-05-3
Chemical Name or Material Palladium
TSCA TSCA
MDL Number MFCD03457879
Physical Form Solid