Please login to your online account to display your discounted pricing

Alfa Aesar™ Palladium, 5% on activated carbon paste, A503023-5

Debenzylation

Chemical Identifiers

Chemical Name or Material Palladium
CAS 7440-05-3
Name Note 5% on activated carbon paste, A503023-5
MDL Number MFCD03457879
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1169406

  SDS
alfa aesar™
1169406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1169414

  SDS
alfa aesar™
1169414
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1169422

  SDS
alfa aesar™
1169422
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Palladium
TSCA TSCA
Name Note 5% on activated carbon paste, A503023-5
MDL Number MFCD03457879
CAS 7440-05-3
Color Black
Quantity 5g
Odor Odorless