missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Palladium, 5% on activated carbon paste, A503023-5

Debenzylation

Manufacturer:  Alfa Aesar™ 1169414

 View more versions of this product

Catalog No. AA1169414



Specifications

Specifications

Palladium
Black
7440-05-3
MFCD03457879
KDLHZDBZIXYQEI-UHFFFAOYSA-N
palladium
23938
25g
Odorless
5% on activated carbon paste, A503023-5
Pd
black
[Pd]
106.42
CHEBI:33363

Chemical Identifiers

7440-05-3
106.42
KDLHZDBZIXYQEI-UHFFFAOYSA-N
23938
palladium
Pd
MFCD03457879
black
CHEBI:33363
[Pd]
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents