missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Palladium, 5% on activated carbon paste, A405032-5

Manufacturer:  Alfa Aesar™ 4549906

 View more versions of this product

Catalog No. AA4549906



Specifications

Specifications

Palladium
7440-05-3
MFCD03457879
KDLHZDBZIXYQEI-UHFFFAOYSA-N
palladium
23938
Odorless
5g
5% on activated carbon paste, A405032-5
Pd
black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon
[Pd]
106.42
CHEBI:33363
Black

Chemical Identifiers

7440-05-3
106.42
KDLHZDBZIXYQEI-UHFFFAOYSA-N
23938
palladium
Pd
MFCD03457879
black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon
CHEBI:33363
[Pd]
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS Documents