missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Palladium, 5% on activated carbon paste, 5R39, Alfa Aesar™

Debenzylation, C-N and C-O cleavage, alkene hydrogenation

Manufacturer:  Thermo Fisher Scientific 4433706

 View more versions of this product

Catalog No. AA4433706



Specifications

Specifications

Palladium
7440-05-3
MFCD03457879
Insoluble in water.
[Pd]
106.42
CHEBI:33363
Black
5% on activated carbon paste, 5R39
Pd
black
KDLHZDBZIXYQEI-UHFFFAOYSA-N
palladium
23938
Odorless
5g

Chemical Identifiers

7440-05-3
106.42
KDLHZDBZIXYQEI-UHFFFAOYSA-N
23938
palladium
Pd
MFCD03457879
black
CHEBI:33363
[Pd]
Safety and Handling

Safety and Handling

EINECSNumber : 231-115-6

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents