missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Palladium, 4.5% Rhodium, 0.5% on carbon paste, F101023-4.5/0.5

Chemical Identifiers

Chemical Name or Material Palladium, 4.5% Rhodium, 0.5% on carbon paste, F101023-4.5
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH3716906 SDS Alfa Aesar™
H3716906
5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAH3716914 SDS Alfa Aesar™
H3716914
25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAH3716922 SDS Alfa Aesar™
H3716922
100g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

Palladium, 4.5% Rhodium, 0.5% on carbon paste, F101023-4.5

Specifications

Solid
Dark gray
Palladium, 4.5% Rhodium, 0.5% on carbon paste, F101023-4.5
Odorless
5g