Please login to your online account to display your discounted pricing

Alfa Aesar™ Palladium, 3% on activated carbon powder, standard, unreduced, nominally 50% water wet

Chemical Identifiers

Chemical Name or Material Palladium
Name Note 3% on activated carbon powder, Standard, unreduced, nominally 50% water wet
MDL Number MFCD03457879
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3829706

 
alfa aesar™
3829706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3829714

 
alfa aesar™
3829714
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3829722

 
alfa aesar™
3829722
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Palladium
TSCA TSCA
Name Note 3% on activated carbon powder, Standard, unreduced, nominally 50% water wet
MDL Number MFCD03457879
Quantity 5g
Odor Odorless
Weight 900 to 1100m2/g