Please login to your online account to display your discounted pricing

Alfa Aesar™ Palladium, 3% on activated carbon powder, eggshell, reduced

Chemical Identifiers

Chemical Name or Material Palladium
Name Note 3% on activated carbon powder, Eggshell, Reduced
MDL Number MFCD03457879
UN Number UN3178
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1169106

  SDS
alfa aesar™
1169106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1169114

  SDS
alfa aesar™
1169114
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1169122

  SDS
alfa aesar™
1169122
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Health Hazard 3 GHS P Statement
P210-P280-P240-P241-P370+P378a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
Use explosion-proof electrical/ventilating/lighting/ equipment.
In case of fire: Use for extinction: CO2, powder or water spray.
Chemical Name or Material Palladium
Quantity 5g
Weight 900 to 1100m2/g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H228
Flammable solid.
Odor Odorless
DOT Information Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLID, INORGANIC, N.O.S.
MDL Number MFCD03457879
UN Number UN3178
TSCA TSCA
Name Note 3% on activated carbon powder, Eggshell, Reduced
Safety and Handling

Warning

GHS P Statement
P210-P280-P240-P241-P370+P378a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
Use explosion-proof electrical/ventilating/lighting/ equipment.
In case of fire: Use for extinction: CO2, powder or water spray.

GHS H Statement
H228
Flammable solid.