Please login to your online account to display your discounted pricing

Alfa Aesar™ Palladium, 20% on activated carbon powder, standard, reduced, nominally 50% water wet

Chemical Identifiers

Chemical Name or Material Palladium
Name Note 20% on activated carbon powder, Standard, reduced, nominally 50% water wet
MDL Number MFCD03457879
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3830804

  SDS
alfa aesar™
3830804
2g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3830809

  SDS
alfa aesar™
3830809
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3830818

  SDS
alfa aesar™
3830818
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA38308MD

  SDS
alfa aesar™
38308MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Palladium
TSCA TSCA
Weight 900 to 1100m2/g
Name Note 20% on activated carbon powder, Standard, reduced, nominally 50% water wet
Quantity 2g
MDL Number MFCD03457879
Odor Odorless