Please login to your online account to display your discounted pricing

Alfa Aesar™ Palladium, 20% on activated carbon powder, eggshell, unreduced, nominally 50% water wet

Debenzylation

Chemical Identifiers

Chemical Name or Material Palladium
Name Note 20% on activated carbon powder, Eggshell, unreduced, nominally 50% water wet
MDL Number MFCD03457879
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3830706

  SDS
alfa aesar™
3830706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3830714

  SDS
alfa aesar™
3830714
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3830722

  SDS
alfa aesar™
3830722
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Palladium
TSCA TSCA
MDL Number MFCD03457879
Weight 900 to 1100m2/g
Odor Odorless
Quantity 5g
Name Note 20% on activated carbon powder, Eggshell, unreduced, nominally 50% water wet