Please login to your online account to display your discounted pricing

Alfa Aesar™ Palladium, 20% on activated carbon powder, eggshell, reduced, nominally 50% water wet

Chemical Identifiers

Chemical Name or Material Palladium
Name Note 20% on activated carbon powder, Eggshell, reduced, nominally 50% water wet
MDL Number MFCD03457879
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3830606

 
alfa aesar™
3830606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3830614

 
alfa aesar™
3830614
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3830622

 
alfa aesar™
3830622
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Palladium
TSCA TSCA
Name Note 20% on activated carbon powder, Eggshell, reduced, nominally 50% water wet
MDL Number MFCD03457879
Quantity 5g
Weight 900 to 1100m2/g