Please login to your online account to display your discounted pricing

Alfa Aesar™ Palladium, 2.5% Platinum, 2.5% on carbon paste, Type 122

Chemical Identifiers

Chemical Name or Material Palladium, 2.5% Platinum
CAS 7440-05-3
Name Note 2.5% on carbon paste, Type 122
MDL Number MFCD03613605
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3654806

  SDS
alfa aesar™
H3654806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH3654814

  SDS
alfa aesar™
H3654814
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH3654822

  SDS
alfa aesar™
H3654822
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Palladium, 2.5% Platinum
TSCA TSCA
Odor Odorless
Name Note 2.5% on carbon paste, Type 122
Physical Form Solid
MDL Number MFCD03613605
CAS 7440-05-3
Color Black
Quantity 5g