missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Palladium 10% on Carbon (wetted with ca. 55% Water), TCI America™

Manufacturer:  TCI America P14915G

 View more versions of this product

Catalog No. P14915G



Specifications

Specifications

Palladium 10% on Carbon (wetted with ca. 55% Water)
7440-05-3
MFCD03457879
[Pd]
106.42
CHEBI:33363
Powder at 20°C
5g
Pd
KDLHZDBZIXYQEI-UHFFFAOYSA-N
palladium
23938
106.42
Black

Chemical Identifiers

7440-05-3
106.42
KDLHZDBZIXYQEI-UHFFFAOYSA-N
CHEBI:33363
[Pd]
Pd
MFCD03457879
23938
palladium
Safety and Handling

Safety and Handling

RTECSNumber : RT3480500

TSCA : Yes

Documents