Please login to your online account to display your discounted pricing

Alfa Aesar™ Palladium, 10% on carbon, Type 487, nominally 50% water wet

Chemical Identifiers

Chemical Name or Material Palladium
Name Note 10% on carbon, Type 487, nominally 50% water wet
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3420009

  SDS
alfa aesar™
H3420009
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH3420018

  SDS
alfa aesar™
H3420018
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Palladium
Name Note 10% on carbon, Type 487, nominally 50% water wet
TSCA TSCA
Physical Form Powder
Color Dark gray
Quantity 10g