missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Palladium, 10% on carbon, Type 487, dry

$45.32 - $146.16

Chemical Identifiers

CAS 7440-05-3
MDL Number MFCD00011167
UN Number UN1325
Merck Index 14,6989
Chemical Name or Material Palladium, 10% on carbon, Type 487, Dry
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1201204 SDS Alfa Aesar™
A1201204
2g Each for $45.32
AAA1201209 SDS Alfa Aesar™
A1201209*
10g Each for $146.16
AAA1201218 SDS Alfa Aesar™
A1201218
50g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Description

Applications
It is a catalyst used in Stille reaction, hydrogenation reactions, Suzuki-Miyaura cross-coupling and Heck-Mizoroki reactions, deoxygenation, oxidation reactions and coupling reactions.

Solubility
Insoluble in water and organic solvents.

Notes
Palladium on carbon should always be handled under an inert atmosphere (preferably argon), and reaction vessels should be flushed with inert gas before the catalyst is added.
Description & Specifications

Chemical Identifiers

7440-05-3
UN1325
Palladium, 10% on carbon, Type 487, Dry
MFCD00011167
14,6989

Specifications

7440-05-3
UN1325
2g
MFCD00011167
14,6989
Palladium, 10% on carbon, Type 487, Dry

Safety and Handling

GHS H Statement
H228
Flammable solid.

GHS P Statement
P210-P280-P240-P241-P370+P378a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
Use explosion-proof electrical/ventilating/lighting/ equipment.
In case of fire: Use for extinction: CO2, powder or water spray.

Warning

DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.

TSCA : TSCA